Methyl-3-(methylamino)-4-nitrobenzenecarboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 3-(methylamino)-4-nitrobenzoate
046155-500MG
046155-1G
046155-5G
101874-430EA
269.23
USD
101874-430
101874-432
101874-434
Methyl-3-(methylamino)-4-nitrobenzenecarboxylate ≥95%
Methyl-3-(methylamino)-4-nitrobenzenecarboxylate
Hazard: Irritant
Formula:
C₉H₁₀N₂O₄ MW: 210.19 g/mol Melting Pt: 135…136 °C |
MDL Number:
MFCD08689705 CAS Number: 251643-13-7 |
Specification Test Results
Melting point | 135-136°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...