1-[(4-Chlorophenyl)sulfonyl]tetrahydro-4(1H)-pyridinone ≥95%
Supplier: Matrix Scientific
Synonyms:
1-[(4-Chlorophenyl)sulphonyl]piperidin-4-one
046154-500MG
046154-1G
046154-5G
101874-424EA
261.54
USD
101874-424
101874-426
101874-428
1-[(4-Chlorophenyl)sulfonyl]tetrahydro-4(1H)-pyridinone ≥95%
1-[(4-Chlorophenyl)sulfonyl]tetrahydro-4(1H)-pyridinone
Hazard: Irritant
Formula:
C₁₁H₁₂ClNO₃S MW: 273.74 g/mol Melting Pt: 158…160 °C |
MDL Number:
MFCD04125155 CAS Number: 156634-92-3 |
Specification Test Results
Melting point | 158-160°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...