1-[(4-Nitrophenyl)sulfonyl]tetrahydro-4(1H)-pyridinone ≥95%

Supplier: Matrix Scientific

Synonyms: 1-[(4-Nitrophenyl)sulphonyl]piperidin-4-one

046153-500MG 046153-1G 046153-5G
101874-418EA 192.31 USD
101874-418 101874-420 101874-422
1-[(4-Nitrophenyl)sulfonyl]tetrahydro-4(1H)-pyridinone ≥95%
1-[(4-Nitrophenyl)sulfonyl]tetrahydro-4(1H)-pyridinone

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₅S
MW: 284.29 g/mol
Melting Pt: 199…201 °C
MDL Number: MFCD08689704
CAS Number: 924869-20-5

Order Now

Specification Test Results

Melting point 199-201°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR