Methyl-4-piperidinobenzenecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-(piperidin-1-yl)benzoate

046152-500MG 046152-1G 046152-5G
101874-412EA 120 USD
101874-412 101874-414 101874-416
Methyl-4-piperidinobenzenecarboxylate ≥95%
Methyl-4-piperidinobenzenecarboxylate

Hazard: Irritant

Formula: C₁₃H₁₇NO₂
MW: 219.29 g/mol
Melting Pt: 94…96 °C
MDL Number: MFCD08444024
CAS Number: 10338-58-6

Order Now

Specification Test Results

Melting point 94-96°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR