Methyl-4-(1-pyrrolidinyl)benzenecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-(pyrrolidin-1-yl)benzoate

046150-500MG 046150-1G 046150-5G
101874-400EA 156.92 USD
101874-400 101874-402 101874-404
Methyl-4-(1-pyrrolidinyl)benzenecarboxylate ≥95%
Methyl-4-(1-pyrrolidinyl)benzenecarboxylate

Hazard: Irritant

Formula: C₁₂H₁₅NO₂
MW: 205.26 g/mol
Melting Pt: 140…142 °C
MDL Number: MFCD00731259
CAS Number: 129414-26-2

Order Now

Specification Test Results

Melting point 140-142°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR