tert-Butyl-4-(chloromethyl)tetrahydro-1(2H)-pyridinecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Boc-4-(chloromethyl)piperidine, 4-(Chloromethyl)piperidine, N-BOC protected

046149-500MG 046149-1G
101874-396EA 118.46 USD
101874-396 101874-398
tert-Butyl-4-(chloromethyl)tetrahydro-1(2H)-pyridinecarboxylate ≥95%
tert-Butyl-4-(chloromethyl)tetrahydro-1(2H)-pyridinecarboxylate

Hazard: Irritant

Formula: C₁₁H₂₀ClNO₂
MW: 233.74 g/mol
Melting Pt: 49…51 °C
MDL Number: MFCD08689676
CAS Number: 479057-79-9

Order Now

Specification Test Results

Melting point 49-51°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR