4-(3-Nitro-2-pyridinyl)benzenecarboxylic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
4-(3-Nitropyridin-2-yl)benzoic acid
046148-500MG
046148-1G
101874-390EA
375.38
USD
101874-390
101874-392
4-(3-Nitro-2-pyridinyl)benzenecarboxylic acid ≥95%
4-(3-Nitro-2-pyridinyl)benzenecarboxylic acid
Hazard: Irritant
Formula:
C₁₂H₈N₂O₄ MW: 244.21 g/mol Melting Pt: 237…239 °C |
MDL Number:
MFCD08444023 CAS Number: 847446-89-3 |
Specification Test Results
Melting point | 237-239°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...