Methyl-4-(1H-pyrazol-1-yl)benzenecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-(1H-pyrazol-1-yl)benzoate

046146-500MG 046146-1G 046146-5G
101874-378EA 181.54 USD
101874-378 101874-380 101874-382
Methyl-4-(1H-pyrazol-1-yl)benzenecarboxylate ≥95%
Methyl-4-(1H-pyrazol-1-yl)benzenecarboxylate

Hazard: Irritant

Formula: C₁₁H₁₀N₂O₂
MW: 202.21 g/mol
Melting Pt: 119…121 °C
MDL Number: MFCD01821225
CAS Number: 400750-29-0

Order Now

Specification Test Results

Melting point 119-121°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR