Ethyl-5-amino-1-benzofuran-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 5-Aminobenzofuran-2-carboxylate, Ethyl 5-aminobenzo[b]furan-2-carboxylate, 5-Aminobenzofuran-2-carboxylic acid ethyl ester

046141-1G 046141-5G 046141-25G
101874-358EA 61.54 USD
101874-358 102551-342 102551-344
Ethyl-5-amino-1-benzofuran-2-carboxylate ≥95%
Ethyl-5-amino-1-benzofuran-2-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₁NO₃
MW: 205.22 g/mol
Melting Pt: 54…56 °C
MDL Number: MFCD08275091
CAS Number: 174775-48-5

Order Now

Specification Test Results

Melting point 54-56°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR