Methyl-5-(4-bromophenoxy)-2-furoate ≥95%

Supplier: Matrix Scientific

046138-500MG 046138-1G 046138-5G
101874-342EA 278.46 USD
101874-342 101874-344 101874-346
Methyl-5-(4-bromophenoxy)-2-furoate ≥95%
Methyl-5-(4-bromophenoxy)-2-furoate

Hazard: Irritant

Formula: C₁₂H₉BrO₄
MW: 297.11 g/mol
Melting Pt: 64…66 °C
MDL Number: MFCD09607928
CAS Number: 952183-41-4

Order Now

Specification Test Results

Melting point 64-66°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR