4'-Chloro-3'-fluoro[1,1'-biphenyl]-2-carbaldehyde ≥95%

Supplier: Matrix Scientific

046134-500MG 046134-1G 046134-5G
101874-320EA 375.38 USD
101874-320 101874-322 101874-324
4'-Chloro-3'-fluoro[1,1'-biphenyl]-2-carbaldehyde ≥95%
4'-Chloro-3'-fluoro[1,1'-biphenyl]-2-carbaldehyde

Hazard: Irritant

Formula: C₁₃H₈ClFO
Melting Pt: 60…62 °C
MDL Number: MFCD08443962
CAS Number: 223575-82-4

Order Now

Specification Test Results

Melting point 60-62°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR