4-(4-Chloro-3-fluorophenyl)benzaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 4'-Chloro-3'-fluoro-[1,1'-biphenyl]-4-carboxaldehyde, 4'-Chloro-3'-fluoro[1,1'-biphenyl]-4-carbaldehyde

046133-500MG 046133-1G 046133-5G
101874-314EA 375.38 USD
101874-314 101874-316 101874-318
4-(4-Chloro-3-fluorophenyl)benzaldehyde ≥95%
4-(4-Chloro-3-fluorophenyl)benzaldehyde

Hazard: Irritant

Formula: C₁₃H₈ClFO
Melting Pt: 112…115 °C
MDL Number: MFCD06201223
CAS Number: 842140-45-8

Order Now

Specification Test Results

Melting point 112-115°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR