tert-Butyl-4-(4-amino-2-fluorophenyl)tetrahydro-1(2H)-pyrazinecarboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
tert-Butyl 4-(4-amino-2-fluorophenyl)piperazine-1-carboxylate, 4-(4-Amino-2-fluorophenyl)piperazine, N1-BOC protected
046131-500MG
046131-1G
046131-5G
101874-302EA
723.08
USD
101874-302
101874-304
101874-306
tert-Butyl-4-(4-amino-2-fluorophenyl)tetrahydro-1(2H)-pyrazinecarboxylate ≥95%
tert-Butyl-4-(4-amino-2-fluorophenyl)tetrahydro-1(2H)-pyrazinecarboxylate
Hazard: Irritant
Formula:
C₁₅H₂₂FN₃O₂ MW: 295.36 g/mol Melting Pt: 110…113 °C |
MDL Number:
MFCD08443960 CAS Number: 154590-35-9 |
Specification Test Results
Melting point | 110-113°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...