5-Bromo-2-(trifluoromethyl)pyrimidine ≥95%
Supplier: Matrix Scientific
046129-250MG
046129-1G
101874-292EA
2408.04
USD
101874-292
101874-294
5-Bromo-2-(trifluoromethyl)pyrimidine ≥95%
5-Bromo-2-(trifluoromethyl)pyrimidine
Hazard: Irritant
Formula:
C₅H₂BrF₃N₂ MW: 226.98 g/mol Melting Pt: 41…42 °C |
MDL Number:
MFCD09754046 CAS Number: 799557-86-1 |
Specification Test Results
Melting point | 41-42°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...