2,3-Dichloro-5-(trifluoromethyl)isonicotinic acid ≥95%
Supplier: Matrix Scientific
046126-500MG
046126-1G
101874-280EA
533.85
USD
101874-280
101874-282
2,3-Dichloro-5-(trifluoromethyl)isonicotinic acid ≥95%
2,3-Dichloro-5-(trifluoromethyl)isonicotinic acid
Hazard: Irritant
Formula:
C₇H₂Cl₂F₃NO₂ Melting Pt: 192…193 °C |
MDL Number:
MFCD12025872 CAS Number: 1135283-33-8 |
Specification Test Results
Melting point | 192-193°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...