4-{[(4-Bromophenyl)sulfanyl]methyl}aniline ≥95%

Supplier: Matrix Scientific

046124-500MG 046124-1G
101874-272EA 375.38 USD
101874-272 101874-274
4-{[(4-Bromophenyl)sulfanyl]methyl}aniline ≥95%
4-{[(4-Bromophenyl)sulfanyl]methyl}aniline

Hazard: Irritant

Formula: C₁₃H₁₂BrNS
Melting Pt: 104…106 °C
MDL Number: MFCD11642554
CAS Number: 54306-14-8

Order Now

Specification Test Results

Melting point 104-106°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR