2-(Chloromethyl)-6-(trifluoromethyl)imidazo[1,2-a]pyridine ≥95%

Supplier: Matrix Scientific

046121-500MG 046121-1G
101874-258EA 538.46 USD
101874-258 101874-260
2-(Chloromethyl)-6-(trifluoromethyl)imidazo[1,2-a]pyridine ≥95%
2-(Chloromethyl)-6-(trifluoromethyl)imidazo[1,2-a]pyridine

Hazard: Irritant

Formula: C₉H₆ClF₃N₂
Melting Pt: 74…75 °C
MDL Number: MFCD09995167
CAS Number: 118000-42-3

Order Now

Specification Test Results

Melting point 74-75°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR