1-(tert-Butyl)-2-methyl-4-propionyl-1H-pyrrole-1,2-dicarboxylate ≥95%
Supplier: Matrix Scientific
046119-500MG
046119-1G
101874-252EA
556.92
USD
101874-252
101874-254
1-(tert-Butyl)-2-methyl-4-propionyl-1H-pyrrole-1,2-dicarboxylate ≥95%
1-(tert-Butyl)-2-methyl-4-propionyl-1H-pyrrole-1,2-dicarboxylate
Hazard: Irritant
Formula:
C₁₄H₁₉NO₅ Melting Pt: 82…84 °C |
MDL Number:
MFCD12025869 CAS Number: 1135283-32-7 |
Specification Test Results
Melting point | 82-84°C |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...