2-Nitro-5-piperidinobenzenecarboxylic acid ≥95%

Supplier: Matrix Scientific

046118-500MG 046118-1G
101874-248EA 276.92 USD
101874-248 101874-250
2-Nitro-5-piperidinobenzenecarboxylic acid ≥95%
2-Nitro-5-piperidinobenzenecarboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₄N₂O₄
Melting Pt: 162…163 °C
MDL Number: MFCD09787496
CAS Number: 118159-39-0

Order Now

Specification Test Results

Melting point 162-163°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR