2-(4-Bromophenyl)-1,3-dioxolane ≥95%
Supplier: Matrix Scientific
046116-500MG
046116-1G
046116-5G
101874-238EA
115.38
USD
101874-238
101874-240
101874-242
2-(4-Bromophenyl)-1,3-dioxolane ≥95%
2-(4-Bromophenyl)-1,3-dioxolane
Hazard: Irritant
Formula:
C₉H₉BrO₂ Melting Pt: 33…35 °C |
MDL Number:
MFCD01318950 CAS Number: 10602-01-4 |
Specification Test Results
Melting point | 33-35°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...