[6-(Trifluoromethyl)-1-benzothiophen-2-yl]methanol ≥95%
Supplier: Matrix Scientific
046114-500MG
046114-1G
101874-228EA
375.38
USD
101874-228
101874-230
[6-(Trifluoromethyl)-1-benzothiophen-2-yl]methanol ≥95%
[6-(Trifluoromethyl)-1-benzothiophen-2-yl]methanol
Hazard: Irritant
Formula:
C₁₀H₇F₃OS MW: 232.23 g/mol Melting Pt: 98…100 °C |
MDL Number:
MFCD11840966 CAS Number: 863118-46-1 |
Specification Test Results
Melting point | 98-100°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...