3-Bromo-6-(trifluoromethyl)imidazo[1,2-a]pyridine ≥95%
Supplier: Matrix Scientific
046109-500MG
046109-1G
046109-5G
101874-208EA
360
USD
101874-208
101874-210
101874-212
3-Bromo-6-(trifluoromethyl)imidazo[1,2-a]pyridine ≥95%
3-Bromo-6-(trifluoromethyl)imidazo[1,2-a]pyridine
Hazard: Irritant
Formula:
C₈H₄BrF₃N₂ Melting Pt: 59…60 °C |
MDL Number:
MFCD12827771 CAS Number: 1146615-86-2 |
Specification Test Results
Melting point | 59-60°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...