6-Fluoroimidazo[1,2-a]pyridine-2-carbohydrazide ≥95%
Supplier: Matrix Scientific
046104-500MG
046104-1G
101874-184EA
921.54
USD
101874-184
101874-186
6-Fluoroimidazo[1,2-a]pyridine-2-carbohydrazide ≥95%
6-Fluoroimidazo[1,2-a]pyridine-2-carbohydrazide
Hazard: Irritant
Formula:
C₈H₇FN₄O Melting Pt: 263…264 °C |
MDL Number:
MFCD11840960 CAS Number: 1135282-88-0 |
Specification Test Results
Melting point | 263-264°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...