(E)-3-(5-Nitro-1-benzofuran-2-yl)-2-propenoic acid ≥95%
Supplier: Matrix Scientific
046103-500MG
046103-1G
101874-180EA
747.69
USD
101874-180
101874-182
(E)-3-(5-Nitro-1-benzofuran-2-yl)-2-propenoic acid ≥95%
(E)-3-(5-Nitro-1-benzofuran-2-yl)-2-propenoic acid
Hazard: Irritant
Formula:
C₁₁H₇NO₅ Melting Pt: 292…294 °C |
MDL Number:
MFCD11840959 CAS Number: 1135282-87-9 |
Specification Test Results
Melting point | 292-294°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...