Methyl-2-(4-methoxy-3-nitrophenyl)acetate ≥95%

Supplier: Matrix Scientific

046101-500MG 046101-1G 046101-5G
101874-170EA 184.62 USD
101874-170 101874-172 101874-174
Methyl-2-(4-methoxy-3-nitrophenyl)acetate ≥95%
Methyl-2-(4-methoxy-3-nitrophenyl)acetate

Hazard: Irritant

Formula: C₁₀H₁₁NO₅
Melting Pt: 100…102 °C
MDL Number: MFCD09754963
CAS Number: 34837-88-2

Order Now

Specification Test Results

Melting point 100-102°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR