2-(4-Chlorophenyl)-4-(trifluoromethyl)-1,3-thiazole-5-carboxylic acid ≥95%
Supplier: Matrix Scientific
046100-500MG
046100-1G
101874-166EA
375.38
USD
101874-166
101874-168
2-(4-Chlorophenyl)-4-(trifluoromethyl)-1,3-thiazole-5-carboxylic acid ≥95%
2-(4-Chlorophenyl)-4-(trifluoromethyl)-1,3-thiazole-5-carboxylic acid
Hazard: Irritant
Formula:
C₁₁H₅ClF₃NO₂S Melting Pt: 180…182 °C |
MDL Number:
MFCD11840957 CAS Number: 187998-87-4 |
Specification Test Results
Melting point | 180-182°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...