2-Morpholino-5-nitroaniline ≥95%

Supplier: Matrix Scientific

046097-500MG 046097-1G
101874-154EA 250.77 USD
101874-154 101874-156
2-Morpholino-5-nitroaniline ≥95%
2-Morpholino-5-nitroaniline

Hazard: Irritant

Formula: C₁₀H₁₃N₃O₃
MW: 223.23 g/mol
Melting Pt: 152…154 °C
MDL Number: MFCD02575506
CAS Number: 4031-79-2

Order Now

Specification Test Results

Melting point 152-154°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR