Methyl-2-chloro-4-(1H-1,2,4-triazol-1-yl)benzenecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-chloro-4-(1H-1,2,4-triazol-1-yl)benzoate

046096-500MG 046096-1G 046096-5G
101874-148EA 275.38 USD
101874-148 101874-150 101874-152
Methyl-2-chloro-4-(1H-1,2,4-triazol-1-yl)benzenecarboxylate ≥95%
Methyl-2-chloro-4-(1H-1,2,4-triazol-1-yl)benzenecarboxylate

Hazard: Irritant

Formula: C₁₀H₈ClN₃O₂
MW: 237.65 g/mol
Melting Pt: 138…140 °C
MDL Number: MFCD09152714
CAS Number: 220141-20-8

Order Now

Specification Test Results

Melting point 138-140°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR