2-(4-Methylphenyl)-2-oxoethyl acetate ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Oxo-2-(p-tolyl)ethyl acetate

046095-500MG 046095-1G 046095-5G
101874-142EA 181.54 USD
101874-142 101874-144 101874-146
2-(4-Methylphenyl)-2-oxoethyl acetate ≥95%
2-(4-Methylphenyl)-2-oxoethyl acetate

Hazard: Irritant

Formula: C₁₁H₁₂O₃
MW: 192.22 g/mol
Melting Pt: 86…88 °C
MDL Number: MFCD00458047
CAS Number: 65143-37-5

Order Now

Specification Test Results

Melting point 86-88°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR