Ethyl-4,6-dichloronicotinate ≥95%
Supplier: Matrix Scientific
046094-1G
046094-5G
046094-25G
101874-138EA
15.38
USD
101874-138
101874-140
102663-726
Ethyl-4,6-dichloronicotinate ≥95%
Ethyl-4,6-dichloronicotinate
Hazard: Irritant/Store in Freezer
Formula:
C₈H₇Cl₂NO₂ Melting Pt: 32…34 °C |
MDL Number:
MFCD00234408 CAS Number: 40296-46-6 |
Specification Test Results
Melting point | 32-34°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...