2-Chloro-N-(2-hydroxyethyl)-4-piperidinobenzenecarboxamide ≥95%
Supplier: Matrix Scientific
046093-500MG
046093-1G
046093-5G
101874-132EA
375.38
USD
101874-132
101874-134
101874-136
2-Chloro-N-(2-hydroxyethyl)-4-piperidinobenzenecarboxamide ≥95%
2-Chloro-N-(2-hydroxyethyl)-4-piperidinobenzenecarboxamide
Hazard: Irritant
Formula:
C₁₄H₁₉ClN₂O₂ Melting Pt: 98…100 °C |
MDL Number:
MFCD09152713 CAS Number: 952183-00-5 |
Specification Test Results
Melting point | 98-100°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...