7-Acetyl-2-methylbenzoxazole ≥95%
Supplier: Matrix Scientific
Synonyms:
1-(2-Methyl-1,3-benzoxazol-7-yl)ethan-1-one
046086-500MG
046086-1G
046086-5G
101874-096EA
278.46
USD
101874-096
101874-098
101874-100
7-Acetyl-2-methylbenzoxazole ≥95%
7-Acetyl-2-methylbenzoxazole
Hazard: Irritant
Formula:
C₁₀H₉NO₂ Melting Pt: 88-100°C |
MDL Number:
MFCD09152710 CAS Number: 952182-98-8 |
Specification Test Results
Melting point | 88-100°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...