1-(1,3-Benzoxazol-7-yl)-3-(dimethylamino)-2-propen-1-one ≥95%
Supplier: Matrix Scientific
046084-500MG
046084-1G
046084-5G
101874-084EA
375.38
USD
101874-084
101874-086
101874-088
1-(1,3-Benzoxazol-7-yl)-3-(dimethylamino)-2-propen-1-one ≥95%
1-(1,3-Benzoxazol-7-yl)-3-(dimethylamino)-2-propen-1-one
Hazard: Irritant
Formula:
C₁₂H₁₂N₂O₂ MW: 216.24 g/mol Melting Pt: 103…105 °C |
MDL Number:
MFCD09152708 CAS Number: 952182-96-6 |
Specification Test Results
Melting point | 103-105°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...