Ethyl-4-(cyclopropylcarbonyl)-3-isoxazolecarboxylate ≥95%
Supplier: Matrix Scientific
046083-500MG
046083-1G
101874-080EA
375.38
USD
101874-080
101874-082
Ethyl-4-(cyclopropylcarbonyl)-3-isoxazolecarboxylate ≥95%
Ethyl-4-(cyclopropylcarbonyl)-3-isoxazolecarboxylate
Hazard: Irritant
Formula:
C₁₀H₁₁NO₄ Melting Pt: 57…59 °C |
MDL Number:
MFCD09152707 CAS Number: 952182-95-5 |
Specification Test Results
Melting point | 57-59°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...