4-(2-Naphthyl)isoxazolo[3,4-d]pyridazin-7(6H)-one ≥95%
Supplier: Matrix Scientific
046078-500MG
046078-1G
046078-5G
101874-056EA
556.92
USD
101874-056
101874-058
101874-060
4-(2-Naphthyl)isoxazolo[3,4-d]pyridazin-7(6H)-one ≥95%
4-(2-Naphthyl)isoxazolo[3,4-d]pyridazin-7(6H)-one
Hazard: Irritant
Formula:
C₁₅H₉N₃O₂ Melting Pt: 260…262 °C |
MDL Number:
MFCD09152703 CAS Number: 952182-91-1 |
Specification Test Results
Melting point | 260-262°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...