1-(5-Methyl-3-phenyl-4-isoxazolyl)-1-ethanone ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Acetyl-5-methyl-3-phenylisoxazole

046076-500MG 046076-1G 046076-5G
101874-046EA 124.62 USD
101874-046 101874-048 101874-050
1-(5-Methyl-3-phenyl-4-isoxazolyl)-1-ethanone ≥95%
1-(5-Methyl-3-phenyl-4-isoxazolyl)-1-ethanone

Hazard: Irritant

Formula: C₁₂H₁₁NO₂
MW: 201.23 g/mol
Melting Pt: 50…52 °C
MDL Number: MFCD00173901
CAS Number: 19212-42-1

Order Now

Specification Test Results

Melting point 50-52°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR