4-(4-Methoxyphenyl)isoxazolo[3,4-d]pyridazin-7(6H)-one ≥95%

Supplier: Matrix Scientific

046074-500MG 046074-1G 046074-5G
101874-034EA 556.92 USD
101874-034 101874-036 101874-038
4-(4-Methoxyphenyl)isoxazolo[3,4-d]pyridazin-7(6H)-one ≥95%
4-(4-Methoxyphenyl)isoxazolo[3,4-d]pyridazin-7(6H)-one

Hazard: Irritant

Formula: C₁₂H₉N₃O₃
Melting Pt: 205…207 °C
MDL Number: MFCD09152700
CAS Number: 952182-88-6

Order Now

Specification Test Results

Melting point 205-207°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR