2-[3-(Trifluoromethoxy)phenoxy]ethanethioamide ≥95%

Supplier: Matrix Scientific

046069-500MG 046069-1G 046069-5G
101874-006EA 556.92 USD
101874-006 101874-008 101874-010
2-[3-(Trifluoromethoxy)phenoxy]ethanethioamide ≥95%
2-[3-(Trifluoromethoxy)phenoxy]ethanethioamide

Hazard: Irritant

Formula: C₉H₈F₃NO₂S
Melting Pt: 90 °C
MDL Number: MFCD09152698
CAS Number: 952182-84-2

Order Now

Specification Test Results

Melting point 90°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR