1-Benzyl-2-(tert-butyl)hydrazine-1,2-dicarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Benzyl 2-(tert-butyl) hydrazine-1,2-dicarboxylate, Hydrazine, N-BOC, N'-CBZ protected

046067-500MG 046067-1G 046067-5G
101873-996EA 61.54 USD
101873-996 101873-998 101874-000
1-Benzyl-2-(tert-butyl)hydrazine-1,2-dicarboxylate ≥95%
1-Benzyl-2-(tert-butyl)hydrazine-1,2-dicarboxylate

Hazard: Irritant

Formula: C₁₃H₁₈N₂O₄
MW: 266.2900 g/mol
Melting Pt: 79…80 °C
MDL Number: MFCD09752862
CAS Number: 57699-88-4

Order Now

Specification Test Results

Melting point 79-80°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR