Benzyl-N-[1,3-benzodioxol-5-yl(phenylsulfonyl)methyl]carbamate ≥95%

Supplier: Matrix Scientific

046065-500MG 046065-1G 046065-5G
101873-984EA 181.54 USD
101873-984 101873-986 101873-988
Benzyl-N-[1,3-benzodioxol-5-yl(phenylsulfonyl)methyl]carbamate ≥95%
Benzyl-N-[1,3-benzodioxol-5-yl(phenylsulfonyl)methyl]carbamate

Hazard: Irritant

Formula: C₂₂H₁₉NO₆S
MW: 425.46 g/mol
Melting Pt: 162…164 °C
MDL Number: MFCD09817425
CAS Number: 952182-82-0

Order Now

Specification Test Results

Melting point 162-164°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR