Ethyl-3-bromo-1H-indole-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 3-bromoindole-2-carboxylate, Ethyl 3-Bromoindole-2-carboxylate, 3-Bromoindole-2-carboxylic acid ethyl ester, Ethyl 3-bromo-1H-indole-2-carboxylate

046063-500MG 046063-1G 046063-5G
101873-972EA 181.54 USD
101873-972 101873-974 101873-976
Ethyl-3-bromo-1H-indole-2-carboxylate ≥95%
Ethyl-3-bromo-1H-indole-2-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₀BrNO₂
MW: 268.11 g/mol
Melting Pt: 149…151 °C
MDL Number: MFCD02071793
CAS Number: 91348-45-7

Order Now

Specification Test Results

Melting point 149-151°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR