3-(3,4-Dimethoxyphenyl)-3-oxopropanenitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 3,4-Dimethoxybenzoylacetonitrile

046058-500MG 046058-1G 046058-5G
101873-954EA 58.46 USD
101873-954 101873-956 101873-958
3-(3,4-Dimethoxyphenyl)-3-oxopropanenitrile ≥95%
3-(3,4-Dimethoxyphenyl)-3-oxopropanenitrile

Hazard: Irritant

Formula: C₁₁H₁₁NO₃
MW: 205.22 g/mol
Melting Pt: 139…141 °C
MDL Number: MFCD02260783
CAS Number: 4640-69-1

Order Now

Specification Test Results

Melting point 139-141°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR