5-Amino-3-(3-chlorophenyl)isoxazole ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(3-Chlorophenyl)isoxazol-5-amine

046057-500MG 046057-1G 046057-5G
101873-948EA 161.54 USD
101873-948 101873-950 101873-952
5-Amino-3-(3-chlorophenyl)isoxazole ≥95%
5-Amino-3-(3-chlorophenyl)isoxazole

Hazard: Irritant

Formula: C₉H₇ClN₂O
Melting Pt: 107-109°C
MDL Number: MFCD07383023
CAS Number: 86685-95-2

Order Now

Specification Test Results

Melting point 107-109°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR