3-(4-Methoxyphenyl)-3-oxopropanenitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Methoxybenzoylacetonitrile

046053-500MG 046053-1G 046053-5G
101873-930EA 53.85 USD
101873-930 101873-932 101873-934
3-(4-Methoxyphenyl)-3-oxopropanenitrile ≥95%
3-(4-Methoxyphenyl)-3-oxopropanenitrile

Hazard: Irritant

Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Melting Pt: 128…130 °C
MDL Number: MFCD00067889
CAS Number: 3672-47-7

Order Now

Specification Test Results

Melting point 128-130°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR