5-Amino-3-(4-chlorophenyl)isoxazole ≥95%
Supplier: Matrix Scientific
Synonyms:
3-(4-Chlorophenyl)isoxazol-5-amine
046052-500MG
046052-1G
046052-5G
101873-924EA
278.46
USD
101873-924
101873-926
101873-928
5-Amino-3-(4-chlorophenyl)isoxazole ≥95%
5-Amino-3-(4-chlorophenyl)isoxazole
Hazard: Irritant
Formula:
C₉H₇ClN₂O MW: 194.6177 g/mol Melting Pt: 162-165°C |
MDL Number:
MFCD01569450 CAS Number: 33866-48-7 |
Specification Test Results
Melting point | 162-165°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...