4-(1,3-Oxazol-5-yl)benzenecarbohydrazide ≥95%

Supplier: Matrix Scientific

046050-500MG 046050-1G
101873-914EA 220 USD
101873-914 101873-916
4-(1,3-Oxazol-5-yl)benzenecarbohydrazide ≥95%
4-(1,3-Oxazol-5-yl)benzenecarbohydrazide

Hazard: Irritant

Formula: C₁₀H₉N₃O₂
MW: 203.2 g/mol
Melting Pt: 208…210 °C
MDL Number: MFCD08276966
CAS Number: 886362-14-7

Order Now

Specification Test Results

Melting point 208-210°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR