2-(5-Methyl-2-phenyl-1,3-thiazol-4-yl)acetic acid ≥95%

Supplier: Matrix Scientific

046049-500MG 046049-1G 046049-5G
101873-908EA 375.38 USD
101873-908 101873-910 101873-912
2-(5-Methyl-2-phenyl-1,3-thiazol-4-yl)acetic acid ≥95%
2-(5-Methyl-2-phenyl-1,3-thiazol-4-yl)acetic acid

Hazard: Irritant

Formula: C₁₂H₁₁NO₂S
MW: 233.29 g/mol
Melting Pt: 135…137 °C
MDL Number: MFCD00277218
CAS Number: 101736-22-5

Order Now

Specification Test Results

Melting point 135-137°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR