Ethyl-2-(2-amino-5-methyl-1,3-thiazol-4-yl)acetate ≥95%

Supplier: Matrix Scientific

046048-500MG 046048-1G 046048-5G
101873-902EA 278.46 USD
101873-902 101873-904 101873-906
Ethyl-2-(2-amino-5-methyl-1,3-thiazol-4-yl)acetate ≥95%
Ethyl-2-(2-amino-5-methyl-1,3-thiazol-4-yl)acetate

Hazard: Irritant

Formula: C₈H₁₂N₂O₂S
MW: 200.26 g/mol
Melting Pt: 116…118 °C
MDL Number: MFCD08443952
CAS Number: 859521-92-9

Order Now

Specification Test Results

Melting point 116-118°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR