Methyl-2,5-difluoro-4-nitrobenzoate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 2,5-difluoro-4-nitrobenzoate
046046-500MG
046046-5G
101873-890EA
103.08
USD
101873-890
101873-894
Methyl-2,5-difluoro-4-nitrobenzoate ≥95%
Methyl 2,5-difluoro-4-nitrobenzoate
Hazard: Irritant
Formula:
C₈H₅F₂NO₄ MW: 217.13 g/mol Melting Pt: 49-52°C |
MDL Number:
MFCD08689670 CAS Number: 924868-81-5 |
Specification Test Results
Melting point | 49-52°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...