2-(4-Benzylpiperazin-1-yl)benzaldehyde ≥95%

Supplier: Matrix Scientific

046043-500MG 046043-1G 046043-5G
101873-878EA 181.54 USD
101873-878 101873-880 101873-882
2-(4-Benzylpiperazin-1-yl)benzaldehyde ≥95%
2-(4-Benzylpiperazin-1-yl)benzaldehyde

Hazard: Irritant

Formula: C₁₈H₂₀N₂O
Melting Pt: 57…59 °C
CAS Number: 112253-26-6

Order Now

Specification Test Results

Melting point 57-59°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR